siRNA Details
Gene Symbol: Lppr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181634.2 | NP_853665.1 | 4 | 878 | ||
GenBank mRNA | AY147866.1 | - | 4 | 878 | ||
AY300027.1 | - | 4 | 878 | |||
BC129083.1 | - | 3 | 714 | |||
BC167750.1 | - | 4 | 679 | 8366697 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
