siRNA Details
Gene Symbol: Adamtsl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240915.2 | XP_006240977.1 | 14 | 2775 | ||
XM_006240916.2 | XP_006240978.1 | 14 | 2765 | |||
XM_006240917.2 | XP_006240979.1 | 15 | 2688 | |||
XM_006240918.2 | XP_006240980.1 | 14 | 2715 | |||
XM_006240919.2 | XP_006240981.1 | 13 | 2575 | |||
XM_006240920.2 | XP_006240982.1 | 13 | 2604 | |||
GenBank mRNA | FQ224249.1 | - | 15 | 2195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
