siRNA Details
Gene Symbol: Tmprss9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047100.2 | NP_001040565.2 | 5 | 505 | ||
XM_006240931.1 | XP_006240993.1 | 7 | 1109 | |||
GenBank mRNA | AB109392.1 | - | 6 | 619 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
