siRNA Details
Gene Symbol: Tmprss9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240931.1 | XP_006240993.1 | 4 | 811 | ||
GenBank mRNA | AB109392.1 | - | 3 | 321 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
