siRNA Details
Gene Symbol: Tcp11l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017458.1 | NP_001017458.1 | 5 | 737 | ||
XM_006241174.2 | XP_006241236.1 | 6 | 799 | |||
GenBank mRNA | BC092644.1 | - | 5 | 737 | 7318345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
