siRNA Details
Gene Symbol: Ikbip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241250.2 | XP_006241312.1 | 3 | 998 | ||
GenBank mRNA | BC088210.1 | - | 3 | 902 | 7382451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
