siRNA Details
Gene Symbol: LOC100911483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277222.1 | NP_001264151.1 | 2 | 144 | ||
GenBank mRNA | FM065403.1 | - | 2 | 140 | ||
FQ211515.1 | - | 2 | 212 | |||
FQ221417.1 | - | 2 | 149 | |||
FQ223566.1 | - | 2 | 155 | |||
FQ223617.1 | - | 2 | 154 | |||
FQ223934.1 | - | 2 | 169 | |||
FQ224478.1 | - | 2 | 157 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
