siRNA Details
Gene Symbol: RGD1307947 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014083.2 | NP_001014105.2 | 5 | 583 | ||
GenBank mRNA | BC079461.1 | - | 5 | 552 | 7122037 | |
CK475399.1 | - | 5 | 570 | |||
CK478081.1 | - | 5 | 551 |
