siRNA Details
Gene Symbol: Osbpl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750319.3 | XP_003750367.1 | 15, 16 | 2065 | ||
XM_006241324.2 | XP_006241386.1 | 14, 15 | 3061 | |||
XM_006241325.1 | XP_006241387.1 | 13, 14 | 1632 | |||
XM_006241326.2 | XP_006241388.1 | 13, 14 | 3024 | |||
XM_008765365.1 | XP_008763587.1 | 15, 16 | 1784 | |||
XM_008765367.1 | XP_008763589.1 | 14, 15 | 8489 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
