siRNA Details
Gene Symbol: Osbpl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750319.3 | XP_003750367.1 | 9 | 1124 | ||
XM_006241324.2 | XP_006241386.1 | 8 | 2120 | |||
XM_006241325.1 | XP_006241387.1 | 7 | 691 | |||
XM_006241326.2 | XP_006241388.1 | 7 | 2083 | |||
XM_008765365.1 | XP_008763587.1 | 9 | 843 | |||
XM_008765367.1 | XP_008763589.1 | 8 | 7548 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
