siRNA Details
Gene Symbol: Frs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108097.3 | NP_001101567.2 | 10 | 1619 | ||
XM_006241381.2 | XP_006241443.1 | 8 | 1561 |

More Information
Other Species Targeted

Gene Ontology Categories:
