siRNA Details
Gene Symbol: Dyrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108100.1 | NP_001101570.1 | 2 | 148 | ||
XM_006241389.2 | XP_006241451.1 | 3 | 377 | |||
XM_006241390.2 | XP_006241452.1 | 3 | 343 | |||
XM_006241391.2 | XP_006241453.1 | 3 | 363 | |||
XM_008765401.1 | XP_008763623.1 | 3 | 753 |
