siRNA Details
Gene Symbol: Dyrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108100.1 | NP_001101570.1 | 2 | 916 | ||
XM_006241389.2 | XP_006241451.1 | 3 | 1145 | |||
XM_006241390.2 | XP_006241452.1 | 3 | 1111 | |||
XM_006241391.2 | XP_006241453.1 | 3 | 1131 | |||
XM_008765401.1 | XP_008763623.1 | 3 | 1521 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001014390, BC085145, AK155496, AK149371.
Gene Ontology Categories:
