siRNA Details
Gene Symbol: Irak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108101.1 | NP_001101571.1 | 2 | 436 | ||
GenBank mRNA | BC166731.1 | - | 2 | 511 | 7934689 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Irak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108101.1 | NP_001101571.1 | 2 | 436 | ||
GenBank mRNA | BC166731.1 | - | 2 | 511 | 7934689 |