siRNA Details
Gene Symbol: Irak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108101.1 | NP_001101571.1 | 2 | 306 | ||
GenBank mRNA | BC166731.1 | - | 2 | 381 | 7934689 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028679, AF461763, AK014783, BC120829, AJ440757, and others.
Gene Ontology Categories:
