siRNA Details
Gene Symbol: Jrk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104612.2 | NP_001098082.1 | 2 | 1306 | ||
XM_006241728.2 | XP_006241790.1 | 2 | 1888 | |||
XM_008765569.1 | XP_008763791.1 | 2 | 1450 | |||
GenBank mRNA | BC152551.1 | - | 1 | 1050 | 7133465 |

More Information
Other Species Targeted

Gene Ontology Categories:
