siRNA Details
Gene Symbol: Smpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729839.4 | XP_002729885.2 | 5 | 1296 | ||
XM_006241911.2 | XP_006241973.1 | 4 | 1463 | |||
XM_008765630.1 | XP_008763852.1 | 5 | 1308 | |||
XM_008765631.1 | XP_008763853.1 | 4 | 1421 | |||
GenBank mRNA | BC166812.1 | - | 4 | 1377 | ||
CB578696.1 | - | 3 | 369 | |||
CB715946.1 | - | 3 | 350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
