siRNA Details
Gene Symbol: Smpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729839.4 | XP_002729885.2 | 3 | 1166 | ||
XM_006241911.2 | XP_006241973.1 | 3 | 1264 | |||
XM_008765630.1 | XP_008763852.1 | 3 | 1178 | |||
XM_008765631.1 | XP_008763853.1 | 3 | 1222 | |||
GenBank mRNA | BC166812.1 | - | 3 | 1178 | ||
CB578696.1 | - | 2 | 329 | |||
CB610411.1 | - | 3 | 437 | |||
CB715946.1 | - | 2 | 310 |

More Information
Other Species Targeted

Gene Ontology Categories:
