siRNA Details
Gene Symbol: Smpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729839.4 | XP_002729885.2 | 1 | 772 | ||
XM_006241911.2 | XP_006241973.1 | 1 | 870 | |||
XM_008765630.1 | XP_008763852.1 | 1 | 772 | |||
XM_008765631.1 | XP_008763853.1 | 1 | 816 | |||
GenBank mRNA | BC166812.1 | - | 1 | 784 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
