siRNA Details
Gene Symbol: Tssk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014086.2 | NP_001014108.2 | 1 | 419 | ||
GenBank mRNA | BC089206.1 | - | 1 | 399 | 7135758 |

More Information
Other Species Targeted

Gene Ontology Categories:
