siRNA Details
Gene Symbol: ARSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001669.3 | NP_001660.2 | 2 | 191 | ||
NM_009589.3 | NP_033667.2 | 2 | 191 | |||
XM_005274514.2 | XP_005274571.1 | 2 | 161 | |||
XM_005274515.2 | XP_005274572.1 | 2 | 161 | |||
GenBank mRNA | AF160499.1 | - | 2 | 91 | ||
AK314479.1 | - | 2 | 155 | |||
BC020229.1 | - | 119 | ||||
BC080179.1 | - | 2 | 94 | |||
DA869167.1 | - | 2 | 191 | |||
X83572.1 | - | 2 | 167 |
