siRNA Details
Gene Symbol: Apol3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013175.1 | NP_001013193.1 | 4 | 758 | ||
NM_001277359.1 | NP_001264288.1 | 4 | 840 | |||
GenBank mRNA | BC086447.1 | - | 4 | 758 | 7304352 | |
FQ233422.1 | - | 4 | 922 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
