siRNA Details
Gene Symbol: Mfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199110.1 | NP_954541.1 | 1 | 174 | ||
GenBank mRNA | BC061801.1 | - | 1 | 174 | 5598335 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
