siRNA Details
Gene Symbol: Card10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130554.1 | NP_001124026.1 | 16 | 2362 | ||
XM_006241986.2 | XP_006242048.1 | 16 | 2488 | |||
XM_006241987.2 | XP_006242049.1 | 16 | 2488 | |||
XM_006241988.2 | XP_006242050.1 | 16 | 3403 |
