siRNA Details
Gene Symbol: Kdelr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127546.1 | NP_001121018.1 | 2 | 186 | ||
XM_008765714.1 | XP_008763936.1 | 2 | 904 | |||
GenBank mRNA | BC162033.1 | - | 2 | 186 | 9103157 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
