siRNA Details
Gene Symbol: Desi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025703.1 | NP_001020874.1 | 3 | 327 | ||
GenBank mRNA | BC098857.1 | - | 3 | 327 | 7380029 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_134095, BC071205, BC132642, BC131979, AK131703, and others.
Gene Ontology Categories:
