siRNA Details
Gene Symbol: Ndufa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130505.1 | NP_001123977.1 | 3 | 310 | ||
GenBank mRNA | FQ218741.1 | - | 3 | 303 | ||
FQ223548.1 | - | 3 | 298 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
