siRNA Details
Gene Symbol: Alg12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108104.2 | NP_001101574.2 | 4 | 379 | ||
GenBank mRNA | BC161934.1 | - | 4 | 377 | 9022144 | |
FM050024.1 | - | 4 | 379 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Alg12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108104.2 | NP_001101574.2 | 4 | 379 | ||
GenBank mRNA | BC161934.1 | - | 4 | 377 | 9022144 | |
FM050024.1 | - | 4 | 379 |