siRNA Details
Gene Symbol: Tymp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012122.1 | NP_001012122.1 | 2 | 213 | ||
XM_006242209.2 | XP_006242271.1 | 2 | 480 | |||
XM_008765730.1 | XP_008763952.1 | 2 | 480 | |||
XM_008765731.1 | XP_008763953.1 | 2 | 480 | |||
XM_008765732.1 | XP_008763954.1 | 2 | 480 | |||
XM_008765733.1 | XP_008763955.1 | 2 | 432 | |||
GenBank mRNA | BC089830.1 | - | 2 | 213 | 7323197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
