siRNA Details
Gene Symbol: Mapk8ip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_235565.8 | XP_235565.5 | 5 | 1893 | ||
GenBank mRNA | BC105884.1 | - | 1 | 816 |

More Information
Other Species Targeted

Gene Ontology Categories:
