siRNA Details
Gene Symbol: Tmem106c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008358.1 | NP_001008359.1 | 8 | 1372 | ||
XM_006242326.2 | XP_006242388.1 | 8 | 1560 | |||
GenBank mRNA | BC086606.1 | - | 8 | 1372 | 7316815 |
