siRNA Details
Gene Symbol: Racgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108112.1 | NP_001101582.1 | 5 | 662 | ||
XM_006257400.2 | XP_006257462.1 | 5 | 609 | |||
GenBank mRNA | BC166473.1 | - | 5 | 562 | 9090730 |
Gene Symbol: LOC103690014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765882.1 | XP_008764104.1 | 3 | 238 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_012025, NM_001253808, NM_001253809, AK200143, AK207314, and others.
Gene Ontology Categories:
