siRNA Details
Gene Symbol: Mfsd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126282.1 | NP_001119754.1 | 2 | 1158 | ||
GenBank mRNA | BC161964.1 | - | 2 | 1158 | 9028283 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_134100, AK087550, BC009140, AK089268, AK165774.
Gene Ontology Categories:
