siRNA Details
Gene Symbol: Atf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108115.1 | NP_001101585.1 | 9 | 926 | ||
XM_006242396.2 | XP_006242458.1 | 9 | 959 | |||
XM_008765746.1 | XP_008763968.1 | 8 | 2617 | |||
XM_008765747.1 | XP_008763969.1 | 9 | 1144 | |||
XM_008765748.1 | XP_008763970.1 | 6 | 2759 | |||
GenBank mRNA | BC158605.1 | - | 1005 | 7135404 |
