siRNA Details
Gene Symbol: Irs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_235721.9 | XP_235721.6 | 1 | 2749 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Irs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_235721.9 | XP_235721.6 | 1 | 2749 |