siRNA Details
Gene Symbol: Tmprss6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130556.1 | NP_001124028.1 | 11 | 1554 | ||
XM_006241995.2 | XP_006242057.1 | 11 | 1571 | |||
XM_006241996.2 | XP_006242058.1 | 11 | 1536 |

More Information
Other Species Targeted

Gene Ontology Categories:
