siRNA Details
Gene Symbol: Kbtbd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108121.1 | NP_001101591.1 | 2 | 1000 | ||
XM_006242474.2 | XP_006242536.1 | 3 | 1099 | |||
XM_006242476.1 | XP_006242538.1 | 3 | 988 | |||
XM_006242477.2 | XP_006242539.1 | 3 | 847 | |||
XM_006242478.2 | XP_006242540.1 | 2 | 854 |
