siRNA Details
Gene Symbol: Cep57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108124.3 | NP_001101594.2 | 9 | 1216 | ||
XM_006242523.2 | XP_006242585.1 | 9 | 1330 | |||
XM_006242524.2 | XP_006242586.1 | 9 | 1512 | |||
GenBank mRNA | BC079415.1 | - | 9 | 1253 | ||
BC168198.1 | - | 9 | 1207 | 7113061 |
