siRNA Details
Gene Symbol: Panx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270548.1 | NP_001257477.1 | 3 | 912 | ||
NM_001270549.1 | NP_001257478.1 | 4 | 1122 | |||
NM_199397.2 | NP_955429.1 | 4 | 1122 | |||
GenBank mRNA | AJ557015.1 | - | 4 | 1122 | ||
GQ499839.1 | - | 3 | 576 | |||
GQ499840.1 | - | 4 | 786 |
