siRNA Details
Gene Symbol: Slc36a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108127.1 | NP_001101597.1 | 10 | 1207 | ||
XM_008765939.1 | XP_008764161.1 | 7 | 892 |

More Information
Other Species Targeted

Gene Ontology Categories:
