siRNA Details
Gene Symbol: RGD1561444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242701.2 | XP_006242763.1 | 2, 3 | 2800 | ||
XM_006242702.2 | XP_006242764.1 | 2, 3 | 2800 | |||
XM_235936.8 | XP_235936.7 | 2, 3 | 2800 |

More Information
Other Species Targeted

