siRNA Details
Gene Symbol: Eepd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014088.3 | NP_001014110.1 | 6 | 1417 | ||
XM_006242726.2 | XP_006242788.1 | 7 | 1851 | |||
XM_006242727.2 | XP_006242789.1 | 6 | 1740 | |||
GenBank mRNA | BC083816.1 | - | 6 | 1417 | 7110630 | |
BC099214.1 | - | 7 | 1809 | 7384643 |
