siRNA Details
Gene Symbol: Igsf9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242739.2 | XP_006242801.1 | 4 | 806 | ||
XM_006242743.2 | XP_006242805.1 | 4 | 806 | |||
XM_006242744.2 | XP_006242806.1 | 4 | 806 | |||
XM_235959.8 | XP_235959.6 | 4 | 806 |
