siRNA Details
Gene Symbol: Nfrkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108133.1 | NP_001101603.1 | 22 | 2850 | ||
XM_006242753.2 | XP_006242815.1 | 22 | 2893 | |||
XM_006242754.2 | XP_006242816.1 | 22 | 3110 | |||
XM_006242755.2 | XP_006242817.1 | 22 | 2890 | |||
XM_006242756.2 | XP_006242818.1 | 22 | 2890 | |||
XM_006242757.2 | XP_006242819.1 | 22 | 2890 | |||
XM_006242758.2 | XP_006242820.1 | 22 | 2887 | |||
XM_006242759.2 | XP_006242821.1 | 22 | 2857 | |||
XM_006242760.2 | XP_006242822.1 | 22 | 2854 | |||
XM_006242761.2 | XP_006242823.1 | 23 | 2803 |
