siRNA Details
Gene Symbol: Kirrel3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048215.1 | NP_001041680.1 | 16 | 2209 | ||
XM_006242782.2 | XP_006242844.1 | 17 | 2339 | |||
GenBank mRNA | DQ902828.1 | - | 16 | 2209 |

More Information
Other Species Targeted

Gene Ontology Categories:
