siRNA Details
Gene Symbol: Pus3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108134.2 | NP_001101604.1 | 2 | 483 | ||
XM_006242785.1 | XP_006242847.1 | 3 | 516 | |||
XM_006242786.2 | XP_006242848.1 | 3 | 1044 | |||
GenBank mRNA | BC087674.1 | - | 2 | 493 | ||
BC158693.1 | - | 2 | 483 | 9023160 |
