siRNA Details
Gene Symbol: Olr1280 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000517.1 | NP_001000517.1 | 1 | 642 | ||
GenBank mRNA | AF091576.1 | - | 1 | 444 |
Gene Symbol: LOC100910500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749190.2 | XP_003749238.1 | 1 | 642 |
Gene Symbol: LOC100910640 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751481.2 | XP_003751529.1 | 1 | 642 |
