siRNA Details
Gene Symbol: Tbcel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014089.1 | NP_001014111.1 | 5 | 700 | ||
XM_006242859.2 | XP_006242921.1 | 5 | 3347 | |||
XM_008766134.1 | XP_008764356.1 | 6 | 3310 | |||
GenBank mRNA | BC087163.1 | - | 5 | 700 | 7135124 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_173038, AK052261, BC139386, BC047462, BC139387, and others.
