siRNA Details
Gene Symbol: TP63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114980.1 | NP_001108452.1 | 11 | 1582 | ||
NM_003722.4 | NP_003713.3 | 13 | 1812 | |||
XM_005247843.3 | XP_005247900.1 | 13 | 2247 | |||
XM_005247844.4 | XP_005247901.1 | 13 | 1975 | |||
XM_011513251.2 | XP_011511553.1 | 14 | 1853 | |||
XM_011513252.2 | XP_011511554.1 | 13 | 1785 | |||
XM_017007387.1 | XP_016862876.1 | 11 | 1578 | |||
GenBank mRNA | AB010153.1 | - | 11 | 1499 | ||
AB016073.1 | - | 13 | 1750 | |||
AB042841.1 | - | 10 | 1186 | |||
AF075430.1 | - | 12 | 1606 | |||
AF075431.1 | - | 11 | 1441 | |||
AF091627.1 | - | 11 | 1566 | |||
AK304127.1 | - | 11 | 1484 | |||
BC039815.1 | - | 13 | 1812 | 5552611 | ||
KU178435.1 | - | 13 | 1723 | |||
KU178436.1 | - | 13 | 1711 | |||
Y16961.1 | - | 13 | 1750 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
