siRNA Details
Gene Symbol: Kmt2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766179.1 | XP_008764401.1 | 22 | 5827 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001081049, AK220457, L17069.
Gene Ontology Categories:
